Assay Details
Gene Symbol: XRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012255.4 | NP_036387.2 | 1 - 2 | 193 | 78 | |
XM_017027723.1 | XP_016883212.1 | 1 - 2 | 122 | 78 | ||
GenBank mRNA | AF064257.1 | - | 1 - 2 | 146 | 78 | |
AK001573.1 | - | 1 - 2 | 163 | 78 | ||
AK314602.1 | - | 1 - 2 | 132 | 78 | ||
AL136841.1 | - | 1 - 2 | 119 | 78 | ||
CF994999.1 | - | 1 - 2 | 200 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000140.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017027723.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA catabolic process, exonucleolytic
DNA-templated transcription, termination
regulation of transcription, DNA-templated
rRNA processing
RNA processing
mRNA processing
RNA catabolic process
spermatogenesis
cell growth
RNA metabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
transcription termination site sequence-specific DNA binding
nuclease activity
5'-3' exoribonuclease activity
protein binding
5'-3' exonuclease activity
poly(A) RNA binding
metal ion binding
DNA catabolic process, exonucleolytic
DNA-templated transcription, termination
regulation of transcription, DNA-templated
rRNA processing
RNA processing
mRNA processing
RNA catabolic process
spermatogenesis
cell growth
RNA metabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
transcription termination site sequence-specific DNA binding
nuclease activity
5'-3' exoribonuclease activity
protein binding
5'-3' exonuclease activity
poly(A) RNA binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001861.hg.1 | ENST00000377191.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001861.hg.1 | ENST00000430571.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001861.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001861.hg.1 | TC20000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20001859.hg.1 | ENST00000377191.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001859.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001859.hg.1 | TC20000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20001860.hg.1 | ENST00000377191.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001860.hg.1 | ENST00000430571.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001860.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001860.hg.1 | TC20000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20001863.hg.1 | ENST00000377191.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001863.hg.1 | ENST00000539513.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001863.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001863.hg.1 | TC20000140.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006899.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147234.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147234.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147234.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147234.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147234.hg.1 | TC2000006899.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147233.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147233.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147233.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147233.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147233.hg.1 | TC2000006899.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147235.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147235.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147235.hg.1 | ENST00000377191.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147235.hg.1 | NM_012255.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147235.hg.1 | TC2000006899.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 175262 | - | - |
