Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000493.hg.1 |
ENST00000296564.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000493.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000493.hg.1 |
TC05000040.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500145390.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500145390.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500145390.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500145390.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500145390.hg.1 |
TC0500006614.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000296564.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000505464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
ENST00000512608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006614.hg.1 |
NM_015325.2 |
Refseq |