This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01070393_m1
See other GPR182 GE Assays ›
Gene Symbol:
GPR182
Gene Name:
G protein-coupled receptor 182
Gene Aliases:
7TMR, ADMR, AM-R, AMR, G10D, gamrh, hrhAMR
Chromosome Location:
Chr.12: 56993895 - 56998441 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01070393_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR182
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011537825.2 XP_011536127.1 2 - 3 2024 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
intracellular signal transduction
adrenomedullin receptor activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000622922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000622922.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0