Assay Details
Gene Symbol: AIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001624.3 | NP_001615.2 | 2 - 3 | 3503 | 131 | |
XM_005266839.3 | XP_005266896.1 | 4 - 5 | 4341 | 131 | ||
XM_017010333.1 | XP_016865822.1 | 2 - 3 | 3728 | 131 | ||
GenBank mRNA | U83115.1 | - | 2 - 3 | 2758 | 131 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000846.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06009987.hg.1 | ENST00000285105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009987.hg.1 | ENST00000369066.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009987.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06009987.hg.1 | TC06000846.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600161912.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600161912.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600161912.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600161912.hg.1 | ENST00000369066.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600161912.hg.1 | ENST00000633556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600161912.hg.1 | NM_001624.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600161912.hg.1 | TC0600008981.hg.1 | Clariom D TC |
