Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000479996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000479996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014364.hg.1 |
ENST00000332298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014364.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014364.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014364.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014364.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014364.hg.1 |
TC20001046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014372.hg.1 |
ENST00000332298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014372.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014372.hg.1 |
TC20001046.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000479996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000479996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009852.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000479996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000479996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168839.hg.1 |
TC2000009852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168851.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168851.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168851.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168851.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168851.hg.1 |
TC2000009852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
ENST00000332298.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
ENST00000395042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
ENST00000493165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
NM_001039467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
NM_005873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168840.hg.1 |
TC2000009852.hg.1 |
Clariom D TC |