This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01069014_m1
See other LINC01258 GE Assays ›
Gene Symbol:
LINC01258
Gene Name:
long intergenic non-protein coding RNA 1258
Gene Aliases:
-
Chromosome Location:
Chr.4: 38420662 - 38523180 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01069014_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01258
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110951.1 6 - 7 1292 80
GenBank mRNA AK057434.1 - 6 - 7 1292 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TBC1 domain family member 1 TBC1D1 TBC,
TBC1
-292.0kb Anti-sense Chr. 4 - 37891084 - 38139175 on NCBI Build GRCh38
Downstream KLF3 antisense RNA 1 KLF3-AS1 162.7kb Sense Chr. 4 - 38612701 - 38664628 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400010438.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay TC0400010438.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171121.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171121.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171121.hg.1 ENST00000512517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171121.hg.1 NR_110951.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171121.hg.1 TC0400010438.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0