This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068515_m1
See other AIM1 GE Assays ›
Gene Symbol:
AIM1
Gene Name:
absent in melanoma 1
Gene Aliases:
CRYBG1, ST4
Chromosome Location:
Chr.6: 106360808 - 106570449 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068515_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001624.3 NP_001615.2 8 - 9 4140 115
XM_005266839.3 XP_005266896.1 10 - 11 4978 115
XM_017010333.1 XP_016865822.1 8 - 9 4365 115
GenBank mRNA AK300443.1 - 2 - 3 175 115
AK316287.1 - 2 - 3 176 115
BC062788.1 - 2 - 3 198 115
U83115.1 - 8 - 9 3395 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008981.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000846.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AIM1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
carbohydrate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06009995.hg.1 ENST00000285105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009995.hg.1 ENST00000369066.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009995.hg.1 ENST00000535438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009995.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06009995.hg.1 TC06000846.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000487681.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay TC0600008981.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 ENST00000369066.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 ENST00000457437.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 ENST00000633556.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 NM_001624.3 Refseq
Affymetrix Human Clariom D Assay PSR0600161919.hg.1 TC0600008981.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0