This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068192_g1
See other SEPT11 GE Assays ›
Gene Symbol:
SEPT11
Gene Name:
septin 11
Gene Aliases:
-
Chromosome Location:
Chr.4: 76949714 - 77040154 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068192_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEPT11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306147.1 NP_001293076.1 3 - 4 435 86
XM_017008400.1 XP_016863889.1 2 - 3 224 86
XM_017008401.1 XP_016863890.1 2 - 3 236 86
XM_017008402.1 XP_016863891.1 2 - 3 236 86
GenBank mRNA AK302700.1 - 3 - 4 312 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400007913.hg.1

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
protein heterooligomerization
cell division
protein binding
GTP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000264893.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000502584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000504637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000505788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000506731.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510515.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000510641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512575.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000512778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513373.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513373.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513373.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513373.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000513697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000515671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000515671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 ENST00000515671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_001306147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007913.hg.1 NM_018243.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0