Assay Details
Gene Symbol: TRPC4AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015638.2 | NP_056453.1 | 5 - 6 | 562 | 90 | |
NM_199368.1 | NP_955400.1 | 5 - 6 | 562 | 90 | ||
XM_011528772.2 | XP_011527074.1 | 5 - 6 | 584 | 90 | ||
XM_017027799.1 | XP_016883288.1 | 5 - 6 | 584 | 90 | ||
XM_017027800.1 | XP_016883289.1 | 5 - 6 | 584 | 90 | ||
XM_017027801.1 | XP_016883290.1 | 5 - 6 | 415 | 90 | ||
GenBank mRNA | AK074106.1 | - | 5 - 6 | 541 | 90 | |
AK298958.1 | - | 4 - 5 | 509 | 90 | ||
AK303476.1 | - | 4 - 5 | 437 | 90 | ||
AK315582.1 | - | 5 - 6 | 554 | 90 | ||
BC013144.1 | - | 5 - 6 | 531 | 4475866 | 90 | |
BI488542.1 | - | 5 - 6 | 543 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000791.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017027800.1,XM_017027801.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20010576.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010576.hg.1 | ENST00000432634.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010576.hg.1 | ENST00000451813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010576.hg.1 | ENST00000541994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010576.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010576.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010576.hg.1 | TC20000791.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162363.hg.1 | TC2000008955.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
