This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01062581_m1
See other CSK GE Assays ›
Gene Symbol:
CSK
Gene Name:
c-src tyrosine kinase
Gene Aliases:
-
Chromosome Location:
Chr.15: 74782064 - 74803198 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01062581_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005254165.4 XP_005254222.1 1 - 2 81 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005254165.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of cytokine production
adaptive immune response
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
epidermal growth factor receptor signaling pathway
central nervous system development
brain development
negative regulation of cell proliferation
negative regulation of low-density lipoprotein particle clearance
cell migration
cell differentiation
T cell costimulation
negative regulation of interleukin-6 production
negative regulation of kinase activity
adherens junction organization
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
negative regulation of Golgi to plasma membrane protein transport
positive regulation of MAP kinase activity
innate immune response
negative regulation of bone resorption
oligodendrocyte differentiation
negative regulation of phagocytosis
T cell receptor signaling pathway
regulation of Fc receptor mediated stimulatory signaling pathway
negative regulation of ERK1 and ERK2 cascade
cellular response to peptide hormone stimulus
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
protein phosphatase binding
protein kinase A catalytic subunit binding
identical protein binding
metal ion binding
proline-rich region binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0