This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061430_m1
See other DDX58 GE Assays ›
Gene Symbol:
DDX58
Gene Name:
DEXD/H-box helicase 58
Gene Aliases:
RIG-I, RIGI, RLR-1, SGMRT2
Chromosome Location:
Chr.9: 32455302 - 32526324 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061430_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDX58
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014314.3 NP_055129.2 11 - 12 1794 101
GenBank mRNA AF038963.1 - 11 - 12 1793 101
AK023661.1 - 4 - 5 469 101
AK125989.1 - 12 - 13 1999 101
AK301678.1 - 10 - 11 1517 101
AK315040.1 - 11 - 12 1665 101
BC132786.1 - 11 - 12 1742 40146808 101
BC136610.1 - 11 - 12 1742 9020600 101
BX647917.1 - 10 - 11 1531 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009825.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000999.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DDX58

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
detection of virus
response to virus
positive regulation of gene expression
regulation of cell migration
negative regulation of type I interferon production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
regulation of type III interferon production
cytoplasmic pattern recognition receptor signaling pathway in response to virus
RIG-I signaling pathway
positive regulation of transcription factor import into nucleus
response to exogenous dsRNA
innate immune response
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
double-stranded DNA binding
double-stranded RNA binding
single-stranded RNA binding
helicase activity
protein binding
ATP binding
zinc ion binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0