This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061150_m1
See other KHDRBS2 GE Assays ›
Gene Symbol:
KHDRBS2
Gene Name:
KH RNA binding domain containing, signal transduction associated 2
Gene Aliases:
SLM-1, SLM1
Chromosome Location:
Chr.6: 61542679 - 62286227 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061150_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KHDRBS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152688.3 NP_689901.2 3 - 4 615 71
XM_011535515.2 XP_011533817.1 3 - 4 578 71
XM_011535516.1 XP_011533818.1 3 - 4 578 71
XM_011535517.1 XP_011533819.1 3 - 4 578 71
XM_011535519.1 XP_011533821.1 3 - 4 578 71
XM_011535520.1 XP_011533822.1 3 - 4 578 71
XM_017010334.1 XP_016865823.1 3 - 4 578 71
XM_017010335.1 XP_016865824.1 3 - 4 578 71
XM_017010336.1 XP_016865825.1 3 - 4 578 71
XM_017010337.1 XP_016865826.1 3 - 4 578 71
XM_017010339.1 XP_016865828.1 3 - 4 578 71
XM_017010340.1 XP_016865829.1 3 - 4 578 71
XM_017010341.1 XP_016865830.1 3 - 4 578 71
XM_017010342.1 XP_016865831.1 3 - 4 578 71
XM_017010343.1 XP_016865832.1 3 - 4 578 71
XM_017010345.1 XP_016865834.1 3 - 4 578 71
XM_017010347.1 XP_016865836.1 3 - 4 501 71
XM_017010348.1 XP_016865837.1 3 - 4 578 71
GenBank mRNA AK095983.1 - 3 - 4 563 71
AK292043.1 - 3 - 4 562 71
BC034043.1 - 3 - 4 615 4796658 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012175.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001842.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KHDRBS2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
protein binding
poly(A) binding
poly(U) RNA binding
SH3 domain binding
SH2 domain binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06022366.hg.1 ENST00000281156.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06022366.hg.1 ENST00000539571.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06022366.hg.1 NM_152688.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06022366.hg.1 TC06001842.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0600183914.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600183914.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay PSR0600183914.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600183914.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay PSR0600183914.hg.1 TC0600012175.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000281156.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 ENST00000511849.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq
Affymetrix Human Clariom D Assay TC0600012175.hg.1 NM_152688.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0