This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061026_m1
See other DIP2C GE Assays ›
Gene Symbol:
DIP2C
Gene Name:
disco interacting protein 2 homolog C
Gene Aliases:
KIAA0934
Chromosome Location:
Chr.10: 274190 - 689710 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061026_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DIP2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014974.2 NP_055789.1 7 - 8 954 88
XM_005252426.3 XP_005252483.1 8 - 9 1113 88
XM_005252427.3 XP_005252484.1 7 - 8 994 88
XM_005252428.4 XP_005252485.1 8 - 9 1164 88
XM_011519428.2 XP_011517730.1 8 - 9 1095 88
XM_011519429.2 XP_011517731.1 8 - 9 1093 88
XM_011519430.2 XP_011517732.1 7 - 8 970 88
XM_011519431.2 XP_011517733.1 7 - 8 970 88
XM_011519432.1 XP_011517734.1 7 - 8 1395 88
GenBank mRNA AB023151.2 - 7 - 8 952 88
AK126063.1 - 8 - 9 1000 88
BC063313.1 - 7 - 8 954 30349267 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009516.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000961.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DIP2C

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
molecular_function
catalytic activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000434695.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000434695.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000434695.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0