This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01057474_m1
See other PADI4 GE Assays ›
Gene Symbol:
PADI4
Gene Name:
peptidyl arginine deiminase 4
Gene Aliases:
PAD, PAD4, PADI5, PDI4, PDI5
Chromosome Location:
Chr.1: 17308195 - 17364004 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01057474_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PADI4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012387.2 NP_036519.2 13 - 14 1584 112
XM_011541150.1 XP_011539452.1 11 - 12 1396 112
XM_011541151.1 XP_011539453.1 11 - 12 1284 112
XM_011541152.1 XP_011539454.1 10 - 11 1116 112
XM_011541157.1 XP_011539459.1 10 - 11 1063 112
GenBank mRNA AB017919.1 - 13 - 14 1582 112
AK290507.1 - 13 - 14 1582 112
AK314839.1 - 13 - 14 1584 112
BC025718.1 - 13 - 14 1568 5228777 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007105.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000232.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PADI4

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
nucleosome assembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular protein modification process
chromatin modification
protein citrullination
stem cell population maintenance
histone H3-R26 citrullination
histone citrullination
innate immune response
protein-arginine deiminase activity
calcium ion binding
protein binding
arginine deiminase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000468945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000468945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000468945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01003719.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01003719.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01003719.hg.1 TC01000232.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01003716.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01003716.hg.1 ENST00000487048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01003716.hg.1 NM_012387.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01003716.hg.1 TC01000232.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100149600.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100149600.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149600.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100149600.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149600.hg.1 TC0100007105.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000375453.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000468945.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000468945.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000468945.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay TC0100007105.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 ENST00000375448.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 ENST00000467001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 ENST00000487048.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 NM_012387.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149597.hg.1 TC0100007105.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0