This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01056913_m1
See other PLA1A GE Assays ›
Gene Symbol:
PLA1A
Gene Name:
phospholipase A1 member A
Gene Aliases:
PS-PLA1, PSPLA1
Chromosome Location:
Chr.3: 119597848 - 119629811 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01056913_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206960.1 NP_001193889.1 7 - 8 941 70
NM_001206961.1 NP_001193890.1 6 - 7 787 70
NM_001293225.1 NP_001280154.1 7 - 8 984 70
NM_015900.3 NP_056984.1 7 - 8 989 70
NR_120610.1 7 - 8 989 70
XM_017006572.1 XP_016862061.1 8 - 9 1021 70
GenBank mRNA AF035268.1 - 7 - 8 954 70
AF035269.1 - 7 - 8 954 70
AK222705.1 - 7 - 8 952 70
AK301880.1 - 6 - 7 760 70
AK313519.1 - 7 - 8 952 70
BC035347.1 - 7 - 8 941 70
BC047703.2 - 7 - 8 893 6061929 70
BX647516.1 - 7 - 8 984 70
U37591.1 - 7 - 8 949 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008484.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000611.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLA1A

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylserine metabolic process
lipid catabolic process
phosphatidylcholine 1-acylhydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0