Assay Details
Gene Symbol: SLC5A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC041454.1 | - | 1 - 2 | 123 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
