Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011954.hg.1 |
ENST00000308191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011954.hg.1 |
ENST00000367787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011954.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011954.hg.1 |
TC06001015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009545.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
ENST00000308191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
ENST00000446774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
ENST00000615259.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
NM_018945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165527.hg.1 |
TC0600009545.hg.1 |
Clariom D TC |