This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01053797_m1
See other SEC31A GE Assays ›
Gene Symbol:
SEC31A
Gene Name:
SEC31 homolog A, COPII coat complex component
Gene Aliases:
ABP125, ABP130, HSPC275, HSPC334, SEC31L1
Chromosome Location:
Chr.4: 82818509 - 82900571 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01053797_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEC31A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300744.2 NP_001287673.1 12 - 13 1704 116
NM_001300745.2 NP_001287674.1 12 - 13 1704 116
NM_016211.4 NP_057295.2 12 - 13 1704 116
GenBank mRNA AB018358.1 - 12 - 13 1560 116
AK125897.1 - 6 - 7 1308 116
BC143492.2 - 12 - 13 1572 9052000 116
CR933696.1 - 12 - 13 1577 116

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SEC31A

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

antigen processing and presentation of peptide antigen via MHC class I
ER to Golgi vesicle-mediated transport
protein transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
IRE1-mediated unfolded protein response
COPII vesicle coating
response to calcium ion
protein binding
calcium-dependent protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0