This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01052642_s1
See other LOC100128840 GE Assays ›
Gene Symbol:
LOC100128840
Gene Name:
uncharacterized LOC100128840
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01052642_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100128840
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK058065.1 - 1 691 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream adhesion G protein-coupled receptor D1 ADGRD1 GPR133,
PGR25
Anti-sense Chr. 12 - 130953907 - 131141469 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 1257 LINC01257 175.0kb Anti-sense Chr. 12 - 131165011 - 131212931 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348474 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P93675 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 41722 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 8910 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348474 - -
Whole Human Genome Oligo Microarray G4112A A_24_P93675 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P93675 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 7967701 - -
Affymetrix Human Gene 1.1 ST Array 7967700 - -
Affymetrix HT HG-U133+ PM Array Plate 1562783_PM_at - -
Human Genome U133 Plus 2.0 Array 1562783_PM_at - -
Human Genome U133 Plus 2.0 Array 1562783_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562783_at11 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0