This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01052349_m1
See other TRAF3IP1 GE Assays ›
Gene Symbol:
TRAF3IP1
Gene Name:
TRAF3 interacting protein 1
Gene Aliases:
IFT54, MIP-T3, MIPT3, SLSN9
Chromosome Location:
Chr.2: 238320489 - 238400900 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01052349_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAF3IP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015650.3 NP_056465.2 7 - 8 1179 143
GenBank mRNA AF242456.1 - 7 - 8 1278 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200011282.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001453.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAF3IP1

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of a polarized epithelium
kidney development
negative regulation of protein phosphorylation
neural tube patterning
embryonic camera-type eye development
negative regulation of protein complex assembly
negative regulation of type I interferon production
negative regulation of interferon-beta production
embryonic heart tube development
post-anal tail morphogenesis
intraciliary transport
cilium assembly
embryonic digit morphogenesis
negative regulation of defense response to virus
regulation of microtubule cytoskeleton organization
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
protein binding
microtubule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02023415.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02023415.hg.1 ENST00000391994.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02023415.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02023415.hg.1 TC02001453.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000391993.7 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000409739.2 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000462122.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 ENST00000483951.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_001139490.1 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay TC0200011282.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay PSR0200181510.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200181510.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay PSR0200181510.hg.1 ENST00000373327.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200181510.hg.1 NM_015650.3 Refseq
Affymetrix Human Clariom D Assay PSR0200181510.hg.1 TC0200011282.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0