This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01047977_m1
See other RUNX2 GE Assays ›
Gene Symbol:
RUNX2
Gene Name:
runt related transcription factor 2
Gene Aliases:
AML3, CBF-alpha-1, CBFA1, CCD, CCD1, CLCD, OSF-2, OSF2, PEA2aA, PEBP2aA
Chromosome Location:
Chr.6: 45328142 - 45664032 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01047977_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RUNX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015051.3 NP_001015051.3 2 - 3 265 111
NM_001024630.3 NP_001019801.3 2 - 3 265 111
XM_011514960.2 XP_011513262.1 2 - 3 440 111
XM_011514961.2 XP_011513263.1 2 - 3 440 111
XM_011514962.2 XP_011513264.1 2 - 3 440 111
XM_011514963.2 XP_011513265.1 2 - 3 440 111
XM_011514964.2 XP_011513266.1 2 - 3 440 111
XM_011514965.2 XP_011513267.1 2 - 3 440 111
XM_017011391.1 XP_016866880.1 2 - 3 475 111
XM_017011393.1 XP_016866882.1 2 - 3 440 111
XM_017011394.1 XP_016866883.1 2 - 3 440 111
XM_017011395.1 XP_016866884.1 2 - 3 440 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008146.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000621.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RUNX2

Panther Classification:

Molecular Function -

Runt transcription factor

Gene Ontology Categories:

Function(s) Process(es)

ossification
osteoblast differentiation
endochondral ossification
osteoblast fate commitment
chondrocyte differentiation
chondrocyte development
osteoblast development
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
hemopoiesis
neuron differentiation
T cell differentiation
BMP signaling pathway
positive regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
regulation of fibroblast growth factor receptor signaling pathway
odontogenesis of dentin-containing tooth
regulation of odontogenesis of dentin-containing tooth
cellular response to fibroblast growth factor stimulus
regulation of cell differentiation
positive regulation of osteoblast differentiation
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
cell maturation
embryonic cranial skeleton morphogenesis
stem cell differentiation
cellular response to BMP stimulus
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
ATP binding
protein domain specific binding
histone deacetylase binding
bHLH transcription factor binding
repressing transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000473041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000473041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0