This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01046602_m1
See other RPS6KA5 GE Assays ›
Gene Symbol:
RPS6KA5
Gene Name:
ribosomal protein S6 kinase A5
Gene Aliases:
MSK1, MSPK1, RLPK
Chromosome Location:
Chr.14: 90868122 - 91060649 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01046602_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS6KA5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322228.1 NP_001309157.1 6 - 7 909 152
NM_001322229.1 NP_001309158.1 7 - 8 1017 152
NM_001322232.1 NP_001309161.1 6 - 7 798 152
NM_001322233.1 NP_001309162.1 7 - 8 1017 152
NM_001322234.1 NP_001309163.1 7 - 8 1054 152
NM_001322235.1 NP_001309164.1 8 - 9 1157 152
NM_001322236.1 NP_001309165.1 6 - 7 933 152
NM_001322237.1 NP_001309166.1 7 - 8 816 152
NM_001322238.1 NP_001309167.1 5 - 6 525 152
NM_004755.3 NP_004746.2 7 - 8 1017 152
NM_182398.2 NP_872198.1 7 - 8 1017 152
XM_017021786.1 XP_016877275.1 8 - 9 1056 152
GenBank mRNA AF074393.1 - 7 - 8 967 152
AF080000.1 - 7 - 8 983 152
AF090421.1 - 7 - 8 966 152
AK295266.1 - 7 - 8 816 152
AK309925.1 - 7 - 8 1041 152
BC017187.2 - 7 - 8 968 3139114 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400009973.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001434.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPS6KA5

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cytokine production
stimulatory C-type lectin receptor signaling pathway
regulation of transcription, DNA-templated
protein phosphorylation
inflammatory response
epidermal growth factor receptor signaling pathway
axon guidance
histone phosphorylation
positive regulation of CREB transcription factor activity
positive regulation of histone phosphorylation
positive regulation of histone acetylation
intracellular signal transduction
histone H3-S10 phosphorylation
histone H3-S28 phosphorylation
histone H2A-S1 phosphorylation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of NF-kappaB transcription factor activity
interleukin-1-mediated signaling pathway
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
histone kinase activity (H3-S10 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000261991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000418736.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000536315.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000554206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556304.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556304.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000556594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 ENST00000614987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_004755.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009973.hg.1 NM_182398.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0