This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01045300_g1
See other G3BP1 GE Assays ›
Gene Symbol:
G3BP1
Gene Name:
G3BP stress granule assembly factor 1
Gene Aliases:
G3BP, HDH-VIII
Chromosome Location:
Chr.5: 151771915 - 151811352 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01045300_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: G3BP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005754.2 NP_005745.1 9 - 10 1123 123
NM_198395.1 NP_938405.1 9 - 10 1092 123
XM_006714749.3 XP_006714812.1 9 - 10 1112 123
XM_006714750.3 XP_006714813.1 9 - 10 1051 123
XM_017008922.1 XP_016864411.1 9 - 10 1398 123
XM_017008923.1 XP_016864412.1 9 - 10 1367 123
GenBank mRNA AK130003.1 - 6 - 7 607 123
AK222606.1 - 9 - 10 1084 123
AK303580.1 - 9 - 10 1180 123
AK312572.1 - 9 - 10 1086 123
BC006997.1 - 9 - 10 1086 3950738 123
BC108278.1 - 9 - 10 1031 6051310 123
BT019384.1 - 8 - 9 952 123
BX647869.1 - 8 - 9 994 123
BX648012.1 - 9 - 10 1098 123
CR536535.1 - 8 - 9 952 123
U32519.1 - 9 - 10 1073 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500009131.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000844.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene G3BP1

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transport
Ras protein signal transduction
DNA duplex unwinding
negative regulation of canonical Wnt signaling pathway
nucleic acid phosphodiester bond hydrolysis
DNA binding
mRNA binding
ATP-dependent DNA helicase activity
ATP-dependent RNA helicase activity
endonuclease activity
protein binding
ATP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000356245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000394123.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000507878.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000517947.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000517947.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000517947.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000517947.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000517947.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000517947.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000517947.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000518726.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000518726.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000518726.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000519832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000519832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000519832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000519832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000519832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000519832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000519832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000520578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000522761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000523519.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000523519.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000523519.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000523519.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000523519.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000523519.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 ENST00000627077.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_005754.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009131.hg.1 NM_198395.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0