This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01044085_g1
See other RYR3 GE Assays ›
Gene Symbol:
RYR3
Gene Name:
ryanodine receptor 3
Gene Aliases:
RYR-3
Chromosome Location:
Chr.15: 33310773 - 33866103 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01044085_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RYR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001036.4 NP_001027.3 61 - 62 8839 89
NM_001243996.2 NP_001230925.1 61 - 62 8839 89
XM_011521880.2 XP_011520182.1 61 - 62 9024 89
XM_017022468.1 XP_016877957.1 61 - 62 9027 89
XM_017022469.1 XP_016877958.1 61 - 62 9024 89
XM_017022470.1 XP_016877959.1 61 - 62 11361 89
XM_017022471.1 XP_016877960.1 61 - 62 9024 89
XM_017022472.1 XP_016877961.1 61 - 62 9024 89
XM_017022473.1 XP_016877962.1 61 - 62 9027 89
XM_017022474.1 XP_016877963.1 61 - 62 8991 89
XM_017022475.1 XP_016877964.1 61 - 62 9027 89
XM_017022476.1 XP_016877965.1 60 - 61 8919 89
XM_017022477.1 XP_016877966.1 61 - 62 9028 89
GenBank mRNA AB001025.1 - 61 - 62 8844 89
AJ001515.1 - 61 - 62 8825 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500006806.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15000229.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RYR3

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
ion transmembrane transport
release of sequestered calcium ion into cytosol
protein homotetramerization
calcium ion transmembrane transport
cellular response to calcium ion
cellular response to magnesium ion
cellular response to caffeine
cellular response to ATP
regulation of cardiac conduction
ryanodine-sensitive calcium-release channel activity
calcium ion binding
calmodulin binding
calcium-release channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15001413.hg.1 ENST00000361728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15001413.hg.1 ENST00000389232.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15001413.hg.1 ENST00000415757.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15001413.hg.1 NM_001036.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15001413.hg.1 NM_001243996.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15001413.hg.1 TC15000229.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 ENST00000389232.8 Ensembl
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 ENST00000415757.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 ENST00000622037.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 NM_001036.4 Refseq
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 NM_001243996.2 Refseq
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 ENST00000389232.8 Ensembl
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 ENST00000415757.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 ENST00000622037.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 NM_001036.4 Refseq
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 NM_001243996.2 Refseq
Affymetrix Human Clariom D Assay PSR1500147314.hg.1 TC1500006806.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0