Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015933.hg.1 |
ENST00000307546.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015933.hg.1 |
ENST00000369611.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015933.hg.1 |
ENST00000369615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015933.hg.1 |
ENST00000369617.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015933.hg.1 |
NM_001142782.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015933.hg.1 |
NM_152900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015933.hg.1 |
TC01000999.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000307546.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000369611.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000369615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000369617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
NM_001142782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
NM_152900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000307546.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000369611.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000369615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
ENST00000369617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
NM_001142782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
NM_152900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168266.hg.1 |
TC0100009474.hg.1 |
Clariom D TC |