This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01042558_m1
See other SPATA6 GE Assays ›
Gene Symbol:
SPATA6
Gene Name:
spermatogenesis associated 6
Gene Aliases:
HASH, SRF-1, SRF1
Chromosome Location:
Chr.1: 48258643 - 48472208 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01042558_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPATA6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286238.1 NP_001273167.1 12 - 13 1432 81
NM_001286239.1 NP_001273168.1 11 - 12 1438 81
NM_019073.3 NP_061946.1 12 - 13 1480 81
XM_006710700.2 XP_006710763.1 10 - 11 1279 81
XM_011541606.1 XP_011539908.1 10 - 11 1189 81
XM_011541607.2 XP_011539909.1 9 - 10 1043 81
XM_011541608.1 XP_011539910.1 11 - 12 1417 81
XM_011541609.2 XP_011539911.1 10 - 11 1125 81
XM_017001510.1 XP_016856999.1 9 - 10 1229 81
XM_017001512.1 XP_016857001.1 13 - 14 1323 81
XM_017001513.1 XP_016857002.1 10 - 11 1087 81
XM_017001514.1 XP_016857003.1 13 - 14 1365 81
GenBank mRNA AK000869.1 - 12 - 13 1445 81
AK301770.1 - 11 - 12 1381 81
BC020660.1 - 12 - 13 1234 4657425 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014127.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002637.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SPATA6

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
cell differentiation
motile cilium assembly
myosin light chain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000371847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000396199.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000463938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000465990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000465990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000465990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000465990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000471975.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000471975.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000471975.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000471975.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000471975.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000471975.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000471975.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000487629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000490850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 ENST00000603831.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286238.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_001286239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014127.hg.1 NM_019073.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0