This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01042486_m1
See other PADI3 GE Assays ›
Gene Symbol:
PADI3
Gene Name:
peptidyl arginine deiminase 3
Gene Aliases:
PAD3, PDI3
Chromosome Location:
Chr.1: 17249079 - 17284233 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01042486_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PADI3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016233.2 NP_057317.2 11 - 12 1349 116
XM_011541571.2 XP_011539873.1 11 - 12 2960 116
XM_017001463.1 XP_016856952.1 8 - 9 1098 116
GenBank mRNA AB026831.1 - 11 - 12 1350 116
BC041592.1 - 11 - 12 1335 5431445 116
BC109091.1 - 11 - 12 1336 40034659 116
BC109092.1 - 11 - 12 1335 40034660 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007103.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000231.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PADI3

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
protein citrullination
protein-arginine deiminase activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01003675.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01003675.hg.1 NM_016233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01003675.hg.1 TC01000231.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay TC0100007103.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149568.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100149568.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149568.hg.1 ENST00000375460.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100149568.hg.1 NM_016233.2 Refseq
Affymetrix Human Clariom D Assay PSR0100149568.hg.1 TC0100007103.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0