Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025257.hg.1 |
ENST00000310823.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025257.hg.1 |
ENST00000497134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025257.hg.1 |
ENST00000538558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025257.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025257.hg.1 |
TC02001558.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184965.hg.1 |
TC0200011688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000310823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000478059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000497134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
ENST00000618923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011688.hg.1 |
NM_003183.5 |
Refseq |