This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01041866_m1
See other RIPK1 GE Assays ›
Gene Symbol:
RIPK1
Gene Name:
receptor interacting serine/threonine kinase 1
Gene Aliases:
RIP, RIP-1, RIP1
Chromosome Location:
Chr.6: 3063888 - 3115187 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01041866_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIPK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317061.1 NP_001303990.1 3 - 4 559 93
NM_003804.4 NP_003795.2 2 - 3 395 93
XM_005249457.3 XP_005249514.2 2 - 3 318 93
XM_006715237.3 XP_006715300.2 2 - 3 318 93
XM_017011405.1 XP_016866894.1 2 - 3 316 93
GenBank mRNA AB208926.1 - 3 - 4 468 93
AK304701.1 - 1 - 2 122 93
AK314176.1 - 2 - 3 395 93
BC126254.1 - 1 - 2 189 8991970 93
BC126256.1 - 1 - 2 189 8991972 93
U50062.1 - 1 - 2 163 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600006653.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000023.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK304701.1,NM_001317061.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RIPK1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
I-kappaB kinase/NF-kappaB signaling
activation of JUN kinase activity
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
positive regulation of necrotic cell death
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
tumor necrosis factor-mediated signaling pathway
response to tumor necrosis factor
TRIF-dependent toll-like receptor signaling pathway
peptidyl-serine autophosphorylation
positive regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
cellular protein catabolic process
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of necroptotic process
T cell apoptotic process
necroptotic process
regulation of ATP:ADP antiporter activity
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
death-inducing signaling complex assembly
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
ripoptosome assembly
necroptotic signaling pathway
ripoptosome assembly involved in necroptotic process
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of hydrogen peroxide-induced cell death
amyloid fibril formation
positive regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
death receptor binding
protein binding
ATP binding
ubiquitin protein ligase binding
protein complex binding
identical protein binding
death domain binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06000167.hg.1 ENST00000259808.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000167.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000167.hg.1 ENST00000541791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000167.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06000167.hg.1 TC06000023.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06000168.hg.1 ENST00000259808.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000168.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000168.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000168.hg.1 ENST00000541791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000168.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06000168.hg.1 TC06000023.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06000169.hg.1 ENST00000259808.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000169.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000169.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000169.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06000169.hg.1 TC06000023.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06000171.hg.1 ENST00000259808.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000171.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000171.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000171.hg.1 ENST00000541791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000171.hg.1 NM_003804.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06000171.hg.1 TC06000023.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 ENST00000479389.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay PSR0600145558.hg.1 TC0600006653.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600145554.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145554.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145554.hg.1 TC0600006653.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600145555.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145555.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600145555.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay PSR0600145555.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145555.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600145555.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay PSR0600145555.hg.1 TC0600006653.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600145556.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145556.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600145556.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay PSR0600145556.hg.1 ENST00000259808.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600145556.hg.1 ENST00000380409.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600145556.hg.1 NM_003804.5 Refseq
Affymetrix Human Clariom D Assay PSR0600145556.hg.1 TC0600006653.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 132795 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0