This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01039916_m1
See other DNER GE Assays ›
Gene Symbol:
DNER
Gene Name:
delta/notch like EGF repeat containing
Gene Aliases:
UNQ26, bet
Chromosome Location:
Chr.2: 229357629 - 229714570 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01039916_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNER
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139072.3 NP_620711.3 6 - 7 1292 66
XM_005246950.2 XP_005247007.1 6 - 7 1279 66
GenBank mRNA AF442487.1 - 6 - 7 1205 66
AK289553.1 - 6 - 7 1250 66
AK304596.1 - 4 - 5 642 66
AK304631.1 - 4 - 5 671 66
AK308576.1 - 1 - 2 57 66
AY358891.1 - 6 - 7 1255 66
BC024766.2 - 2 - 3 298 66
BC035009.1 - 6 - 7 1282 4820343 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200015958.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002847.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DNER

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
endocytosis
Notch signaling pathway
Notch receptor processing
synapse assembly
central nervous system development
glial cell differentiation
skeletal muscle fiber development
transmembrane signaling receptor activity
Notch binding
calcium ion binding
protein binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02045929.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02045929.hg.1 ENST00000543700.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02045929.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02045929.hg.1 TC02002847.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000482831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000482831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 ENST00000482831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay PSR0200217363.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200217363.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay PSR0200217363.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200217363.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay PSR0200217363.hg.1 TC0200015958.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000341772.4 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000482831.1 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000482831.1 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 ENST00000482831.1 Ensembl
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq
Affymetrix Human Clariom D Assay TC0200015958.hg.1 NM_139072.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0