This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01039417_m1
See other SLF2 GE Assays ›
Gene Symbol:
SLF2
Gene Name:
SMC5-SMC6 complex localization factor 2
Gene Aliases:
C10orf6, FAM178A
Chromosome Location:
Chr.10: 100910230 - 100965134 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01039417_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136123.1 NP_001129595.1 10 - 11 3055 124
NM_018121.3 NP_060591.3 10 - 11 3055 124
XM_005269965.2 XP_005270022.1 10 - 11 2670 124
XM_011539944.2 XP_011538246.1 10 - 11 5645 124
GenBank mRNA AF460991.1 - 10 - 11 3055 124
AK001374.1 - 5 - 6 551 124
AK292565.1 - 10 - 11 2672 124
AL832383.1 - 10 - 11 2622 124
BC030565.1 - 10 - 11 2645 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008661.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000726.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLF2

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
cellular response to DNA damage stimulus
positive regulation of protein complex assembly
positive regulation of maintenance of mitotic sister chromatid cohesion
protein localization to site of double-strand break
positive regulation of double-strand break repair
protein binding
ubiquitin protein ligase binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10008486.hg.1 ENST00000238961.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008486.hg.1 ENST00000370269.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008486.hg.1 NM_001136123.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10008486.hg.1 NM_018121.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10008486.hg.1 TC10000726.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 ENST00000238961.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 ENST00000370269.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 NM_001136123.1 Refseq
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 NM_018121.3 Refseq
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 ENST00000238961.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 ENST00000370269.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 NM_001136123.1 Refseq
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 NM_018121.3 Refseq
Affymetrix Human Clariom D Assay PSR1000159083.hg.1 TC1000008661.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0