This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01038401_m1
See other ENPP3 GE Assays ›
Gene Symbol:
ENPP3
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 3
Gene Aliases:
B10, CD203c, NPP3, PD-IBETA, PDNP3
Chromosome Location:
Chr.6: 131637235 - 131747413 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01038401_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ENPP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005021.4 NP_005012.2 19 - 20 1947 84
NR_133007.1 19 - 20 1947 84
XM_011535897.1 XP_011534199.1 17 - 18 1772 84
XM_017010932.1 XP_016866421.1 17 - 18 1992 84
GenBank mRNA AK024899.1 - 19 - 20 1880 84
AK226107.1 - 20 - 21 2125 84
AK292903.1 - 19 - 20 1878 84
DQ234347.1 - 19 - 20 1797 84
EF560735.1 - 19 - 20 1857 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ENPP3

Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
receptor-mediated endocytosis
immune response
nucleoside triphosphate catabolic process
inorganic diphosphate transport
nucleic acid phosphodiester bond hydrolysis
nucleic acid binding
phosphodiesterase I activity
nucleotide diphosphatase activity
scavenger receptor activity
polysaccharide binding
NADH pyrophosphatase activity
metal ion binding
nucleoside-triphosphate diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header