Assay Details
Gene Symbol: PDK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002612.3 | NP_002603.1 | 1 - 2 | 455 | 91 | |
GenBank mRNA | AK096428.1 | - | 2 - 3 | 254 | 91 | |
AK223328.1 | - | 1 - 2 | 351 | 91 | ||
AK312424.1 | - | 1 - 2 | 361 | 91 | ||
BC040239.1 | - | 1 - 2 | 335 | 3047987 | 91 | |
DB035430.1 | - | 1 - 2 | 457 | 91 | ||
U54617.1 | - | 1 - 2 | 357 | 91 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001618.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK096428.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
glucose metabolic process
protein phosphorylation
regulation of pH
insulin receptor signaling pathway
cellular response to starvation
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of cellular ketone metabolic process
regulation of glucose metabolic process
regulation of fatty acid biosynthetic process
glucose homeostasis
response to starvation
regulation of bone resorption
regulation of fatty acid oxidation
cellular response to fatty acid
reactive oxygen species metabolic process
xenophagy
negative regulation of anoikis
protein kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
ATP binding
positive regulation of defense response to virus by host
glucose metabolic process
protein phosphorylation
regulation of pH
insulin receptor signaling pathway
cellular response to starvation
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of cellular ketone metabolic process
regulation of glucose metabolic process
regulation of fatty acid biosynthetic process
glucose homeostasis
response to starvation
regulation of bone resorption
regulation of fatty acid oxidation
cellular response to fatty acid
reactive oxygen species metabolic process
xenophagy
negative regulation of anoikis
protein kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
ATP binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07024641.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024641.hg.1 | ENST00000542888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024641.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07024641.hg.1 | TC07001618.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07024638.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024638.hg.1 | ENST00000542888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024638.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07024638.hg.1 | TC07001618.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07024639.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024639.hg.1 | ENST00000542888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024639.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07024639.hg.1 | TC07001618.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07024640.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024640.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07024640.hg.1 | TC07001618.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07024646.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07024646.hg.1 | TC07001618.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000468445.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000473796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | ENST00000498190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011842.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184671.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184671.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184671.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184671.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184671.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184671.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184671.hg.1 | TC0700011842.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700184672.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184672.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184672.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184672.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184672.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184672.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184672.hg.1 | TC0700011842.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700184674.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184674.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184674.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184674.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184674.hg.1 | TC0700011842.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700184673.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184673.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184673.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184673.hg.1 | ENST00000005178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184673.hg.1 | ENST00000473301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184673.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184673.hg.1 | TC0700011842.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700184675.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184675.hg.1 | NM_002612.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184675.hg.1 | TC0700011842.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 101060 | - | - |
