This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037471_m1
See other LCP1 GE Assays ›
Gene Symbol:
LCP1
Gene Name:
lymphocyte cytosolic protein 1
Gene Aliases:
CP64, HEL-S-37, L-PLASTIN, LC64P, LPL, PLS2
Chromosome Location:
Chr.13: 46125923 - 46182324 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037471_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LCP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002298.4 NP_002289.2 9 - 10 1214 66
XM_005266374.1 XP_005266431.1 9 - 10 1112 66
GenBank mRNA AB208984.1 - 9 - 10 1059 66
AK223305.1 - 9 - 10 1149 66
AK312393.1 - 9 - 10 1080 66
BC007673.1 - 9 - 10 1052 3143290 66
BC010271.1 - 9 - 10 1116 2900752 66
CR936795.1 - 9 - 10 1102 66
EU794648.1 - 9 - 10 1052 66
J02923.1 - 9 - 10 1050 66
M22300.1 - 7 - 8 805 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LCP1

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

T cell activation involved in immune response
protein kinase A signaling
cell migration
extracellular matrix disassembly
organ regeneration
regulation of intracellular protein transport
wound healing, spreading of cells
actin filament bundle assembly
positive regulation of podosome assembly
actin binding
calcium ion binding
identical protein binding
actin filament binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header