This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037111_m1
See other CABP1 GE Assays ›
Gene Symbol:
CABP1
Gene Name:
calcium binding protein 1
Gene Aliases:
CALBRAIN, HCALB_BR
Chromosome Location:
Chr.12: 120640552 - 120680869 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037111_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CABP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033677.1 NP_001028849.1 1 - 2 720 97
XM_017020235.1 XP_016875724.1 1 - 2 787 97
GenBank mRNA AL533429.3 - 1 - 2 689 97
BC030201.1 - 1 - 2 720 5200625 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200009136.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000948.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CABP1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein import into nucleus
negative regulation of catalytic activity
enzyme inhibitor activity
calcium ion binding
protein binding
nuclear localization sequence binding
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012295.hg.1 ENST00000316803.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012295.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012295.hg.1 TC12000948.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 ENST00000288616.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 ENST00000316803.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 ENST00000351200.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 NM_001033677.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 NM_004276.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 NM_031205.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012303.hg.1 TC12000948.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167564.hg.1 TC1200009136.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200167556.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167556.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay PSR1200167556.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167556.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay PSR1200167556.hg.1 TC1200009136.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200167557.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167557.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay PSR1200167557.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200167557.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay PSR1200167557.hg.1 TC1200009136.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000288616.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000316803.7 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000351200.6 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000453000.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 ENST00000498082.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_001033677.1 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_004276.4 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq
Affymetrix Human Clariom D Assay TC1200009136.hg.1 NM_031205.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0