This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037003_g1
See other MMP14 GE Assays ›
Gene Symbol:
MMP14
Gene Name:
matrix metallopeptidase 14
Gene Aliases:
MMP-14, MMP-X1, MT-MMP, MT-MMP 1, MT1-MMP, MT1MMP, MTMMP1, WNCHRS
Chromosome Location:
Chr.14: 22836533 - 22847600 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037003_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MMP14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004995.3 NP_004986.1 2 - 3 536 77
GenBank mRNA AK291325.1 - 2 - 3 478 77
AK312657.1 - 2 - 3 484 77
BC064803.1 - 2 - 3 470 6046773 77
D26512.1 - 2 - 3 362 77
HE978355.1 - 2 - 3 485 77
U41078.1 - 2 - 3 251 77
X83535.1 - 2 - 3 370 77
X90925.1 - 2 - 3 363 77
Z48481.1 - 2 - 3 364 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400006659.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000133.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MMP14

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
ovarian follicle development
response to hypoxia
endothelial cell proliferation
endochondral ossification
proteolysis
response to oxidative stress
male gonad development
response to mechanical stimulus
response to hormone
positive regulation of myotube differentiation
positive regulation of peptidase activity
response to organic cyclic compound
extracellular matrix disassembly
positive regulation of cell growth
lung development
positive regulation of cell migration
collagen catabolic process
zymogen activation
endodermal cell differentiation
chondrocyte proliferation
astrocyte cell migration
response to estrogen
positive regulation of B cell differentiation
negative regulation of Notch signaling pathway
embryonic cranial skeleton morphogenesis
branching morphogenesis of an epithelial tube
tissue remodeling
negative regulation of focal adhesion assembly
craniofacial suture morphogenesis
metalloendopeptidase activity
serine-type endopeptidase activity
integrin binding
calcium ion binding
protein binding
zinc ion binding
peptidase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0