This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01035542_m1
See other SF3B3 GE Assays ›
Gene Symbol:
SF3B3
Gene Name:
splicing factor 3b subunit 3
Gene Aliases:
RSE1, SAP130, SF3b130, STAF130
Chromosome Location:
Chr.16: 70523788 - 70577668 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01035542_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SF3B3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012426.4 NP_036558.3 6 - 7 1034 86
GenBank mRNA AJ001443.1 - 6 - 7 1060 86
AK291768.1 - 6 - 7 1032 86
BC068974.1 - 6 - 7 971 6168677 86
D87686.1 - 6 - 7 979 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008275.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000579.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SF3B3

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

RNA splicing, via transesterification reactions
mRNA splicing, via spliceosome
mRNA processing
protein complex assembly
RNA splicing
negative regulation of protein catabolic process
RNA binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000564899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000564899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000564899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000569687.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000569687.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000569687.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header