This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01034853_m1
See other HIP1 GE Assays ›
Gene Symbol:
HIP1
Gene Name:
huntingtin interacting protein 1
Gene Aliases:
HIP-I, ILWEQ, SHON, SHONbeta, SHONgamma
Chromosome Location:
Chr.7: 75533298 - 75738976 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01034853_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HIP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005250305.2 XP_005250362.1 1 - 2 176 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005250305.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cytoskeleton organization
cell differentiation
regulation of apoptotic process
positive regulation of receptor-mediated endocytosis
clathrin coat assembly
clathrin-mediated endocytosis
apoptotic signaling pathway
actin binding
structural constituent of cytoskeleton
protein binding
clathrin binding
phosphatidylinositol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0