This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01034257_m1
See other TP53 GE Assays ›
Gene Symbol:
TP53
Gene Name:
tumor protein p53
Gene Aliases:
BCC7, LFS1, P53, TRP53
Chromosome Location:
Chr.17: 7668402 - 7687550 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01034257_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TP53
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000546.5 NP_000537.3 9 - 10 1199 83
NM_001126112.2 NP_001119584.1 9 - 10 1196 83
NM_001126115.1 NP_001119587.1 5 - 6 879 83
NM_001126118.1 NP_001119590.1 8 - 9 1316 83
NM_001276697.1 NP_001263626.1 5 - 6 879 83
NM_001276760.1 NP_001263689.1 9 - 10 1199 83
NM_001276761.1 NP_001263690.1 9 - 10 1196 83
GenBank mRNA AB082923.1 - 9 - 10 1060 83
AF307851.1 - 9 - 10 1132 83
AK223026.1 - 3 - 4 292 83
AK225838.1 - 9 - 10 1137 83
AK297462.1 - 10 - 11 1064 83
AK297927.1 - 10 - 11 1109 83
AK312568.1 - 9 - 10 1141 83
AM076970.1 - 8 - 9 964 83
AY429684.1 - 8 - 9 997 83
BC003596.1 - 9 - 10 1117 3544714 83
BT019622.1 - 8 - 9 997 83
DQ186650.1 - 5 - 6 875 83
DQ191317.1 - 9 - 10 1186 83
DQ263704.1 - 8 - 9 997 83
DQ286964.1 - 9 - 10 1131 83
EF101867.1 - 8 - 9 964 83
FJ207420.1 - 9 - 10 1132 83
JN214348.1 - 8 - 9 1042 83
JN601513.1 - 8 - 9 997 83
JN601514.1 - 8 - 9 997 83
JN601515.1 - 8 - 9 997 83
JN601516.1 - 9 - 10 971 83
JN900492.1 - 8 - 9 1305 83
JQ026207.1 - 8 - 9 987 83
JQ026208.1 - 8 - 9 992 83
JQ026209.1 - 8 - 9 1002 83
JQ026210.1 - 8 - 9 992 83
JQ694049.1 - 8 - 9 1020 83
JQ694050.1 - 8 - 9 1020 83
JQ694051.1 - 8 - 9 1020 83
M14694.1 - 9 - 10 1122 83
M14695.1 - 9 - 10 1118 83
X01405.1 - 6 - 7 698 83
X02469.1 - 9 - 10 1132 83
X60011.1 - 8 - 9 997 83
X60012.1 - 8 - 9 997 83
X60013.1 - 8 - 9 997 83
X60014.1 - 8 - 9 997 83
X60015.1 - 8 - 9 997 83
X60016.1 - 8 - 9 997 83
X60017.1 - 8 - 9 997 83
X60018.1 - 8 - 9 997 83
X60019.1 - 8 - 9 997 83
X60020.1 - 8 - 9 997 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TP53

Panther Classification:

Molecular Function -

P53-like transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA strand renaturation
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
ER overload response
cell cycle arrest
Ras protein signal transduction
multicellular organism development
cell aging
protein localization
cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
response to X-ray
response to gamma radiation
positive regulation of gene expression
viral process
protein sumoylation
cell differentiation
negative regulation of cell growth
DNA damage response, signal transduction by p53 class mediator
positive regulation of histone deacetylation
chromatin assembly
mitotic G1 DNA damage checkpoint
positive regulation of protein oligomerization
cellular response to UV
cellular response to drug
cellular response to glucose starvation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
positive regulation of protein export from nucleus
regulation of mitochondrial membrane permeability
negative regulation of fibroblast proliferation
circadian behavior
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of helicase activity
protein tetramerization
negative regulation of telomerase activity
positive regulation of thymocyte apoptotic process
positive regulation of cell cycle arrest
cellular response to hypoxia
cellular response to ionizing radiation
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
replicative senescence
oxidative stress-induced premature senescence
intrinsic apoptotic signaling pathway
oligodendrocyte apoptotic process
positive regulation of execution phase of apoptosis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
regulation of cell cycle G2/M phase transition
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of reactive oxygen species metabolic process
positive regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protease binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
copper ion binding
protein binding
ATP binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
protein phosphatase binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
identical protein binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
chaperone binding
protein phosphatase 2A binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0