Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034466.hg.1 |
ENST00000289228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034466.hg.1 |
ENST00000460427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034466.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034466.hg.1 |
TC02002121.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034465.hg.1 |
ENST00000289228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034465.hg.1 |
ENST00000460427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034465.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034465.hg.1 |
TC02002121.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199448.hg.1 |
TC0200013573.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199447.hg.1 |
TC0200013573.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000289228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000451664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
ENST00000460427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013573.hg.1 |
NM_005735.3 |
Refseq |