This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01031983_g1
See other WISP2 GE Assays ›
Gene Symbol:
WISP2
Gene Name:
WNT1 inducible signaling pathway protein 2
Gene Aliases:
CCN5, CT58, CTGF-L
Chromosome Location:
Chr.20: 44714204 - 44728041 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01031983_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WISP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001323370.1 NP_001310299.1 1 - 2 320 68
GenBank mRNA BI823598.1 - 1 - 2 318 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001323370.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
cell adhesion
signal transduction
cell-cell signaling
negative regulation of cell death
integrin binding
insulin-like growth factor binding
heparin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000190983.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372865.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000372868.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000465000.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000471629.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 ENST00000497421.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq
Affymetrix Human Clariom D Assay TC2000007465.hg.1 NM_003881.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0