This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01031768_m1
See other UNC5C GE Assays ›
Gene Symbol:
UNC5C
Gene Name:
unc-5 netrin receptor C
Gene Aliases:
UNC5H3
Chromosome Location:
Chr.4: 95162504 - 95549210 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01031768_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC5C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003728.3 NP_003719.3 13 - 14 2641 101
XM_005263321.3 XP_005263378.1 14 - 15 2698 101
XM_017008789.1 XP_016864278.1 14 - 15 9058 101
XM_017008790.1 XP_016864279.1 13 - 14 2286 101
GenBank mRNA AF055634.1 - 13 - 14 2442 101
AK290500.1 - 13 - 14 2452 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011359.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001395.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UNC5C

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
axon guidance
brain development
regulation of cell migration
anterior/posterior axon guidance
netrin-activated signaling pathway
positive regulation of apoptotic process
netrin receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04019417.hg.1 ENST00000331502.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04019417.hg.1 ENST00000453304.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04019417.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04019417.hg.1 TC04001395.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000504962.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000506749.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000513796.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay TC0400011359.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay PSR0400177637.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400177637.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400177637.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay PSR0400177637.hg.1 ENST00000453304.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400177637.hg.1 ENST00000610318.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400177637.hg.1 NM_003728.3 Refseq
Affymetrix Human Clariom D Assay PSR0400177637.hg.1 TC0400011359.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0