Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000581999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008867.hg.1 |
NR_037409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010805.hg.1 |
ENST00000262613.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010805.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010805.hg.1 |
TC17000834.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000581356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000581999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
ENST00000583369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008867.hg.1 |
NR_037409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
ENST00000262613.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
ENST00000413388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
ENST00000578958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
NM_004252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166246.hg.1 |
TC1700008867.hg.1 |
Clariom D TC |