Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008537.hg.1 |
ENST00000357337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008537.hg.1 |
ENST00000407578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008537.hg.1 |
ENST00000544440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008537.hg.1 |
NM_015057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008537.hg.1 |
TC13000744.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
ENST00000357337.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
ENST00000429715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
ENST00000544440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
NM_015057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
ENST00000357337.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
ENST00000429715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
ENST00000544440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
NM_015057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162367.hg.1 |
TC1300009293.hg.1 |
Clariom D TC |