This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01031386_m1
See other MYCBP2 GE Assays ›
Gene Symbol:
MYCBP2
Gene Name:
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases:
Myc-bp2, PAM, Phr
Chromosome Location:
Chr.13: 77044657 - 77327066 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01031386_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYCBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015057.4 NP_055872.4 61 - 62 10992 68
XM_005266299.2 XP_005266356.1 63 - 64 11247 68
XM_006719779.2 XP_006719842.1 64 - 65 11373 68
XM_006719780.2 XP_006719843.1 64 - 65 11373 68
XM_006719781.2 XP_006719844.1 64 - 65 11349 68
XM_006719782.2 XP_006719845.1 63 - 64 11298 68
XM_006719783.2 XP_006719846.1 63 - 64 11271 68
XM_006719784.2 XP_006719847.1 63 - 64 11265 68
XM_006719785.2 XP_006719848.1 63 - 64 11193 68
XM_006719786.2 XP_006719849.1 62 - 63 11091 68
XM_006719787.2 XP_006719850.1 62 - 63 11073 68
XM_006719788.2 XP_006719851.1 62 - 63 11049 68
XM_006719789.2 XP_006719852.1 63 - 64 11247 68
XM_011535004.2 XP_011533306.1 63 - 64 10912 68
XM_011535005.2 XP_011533307.1 47 - 48 8453 68
XM_017020454.1 XP_016875943.1 64 - 65 11316 68
XM_017020455.1 XP_016875944.1 63 - 64 11241 68
XM_017020456.1 XP_016875945.1 63 - 64 11190 68
XM_017020457.1 XP_016875946.1 62 - 63 11172 68
XM_017020458.1 XP_016875947.1 62 - 63 11139 68
XM_017020459.1 XP_016875948.1 61 - 62 10983 68
XM_017020460.1 XP_016875949.1 61 - 62 10971 68
XM_017020461.1 XP_016875950.1 61 - 62 10959 68
XM_017020462.1 XP_016875951.1 61 - 62 10947 68
XM_017020463.1 XP_016875952.1 60 - 61 10908 68
XM_017020464.1 XP_016875953.1 61 - 62 10893 68
XM_017020465.1 XP_016875954.1 60 - 61 10839 68
XM_017020466.1 XP_016875955.1 60 - 61 10791 68
XM_017020467.1 XP_016875956.1 60 - 61 10791 68
XM_017020468.1 XP_016875957.1 63 - 64 10860 68
GenBank mRNA AB020723.1 - 2 - 3 322 68
AF075587.1 - 62 - 63 10760 68
BX647202.1 - 4 - 5 664 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009293.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000744.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MYCBP2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
protein ubiquitination
protein binding
zinc ion binding
ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13008546.hg.1 ENST00000357337.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008546.hg.1 ENST00000407578.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008546.hg.1 ENST00000544440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008546.hg.1 NM_015057.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13008546.hg.1 TC13000744.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300162376.hg.1 ENST00000357337.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300162376.hg.1 ENST00000544440.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300162376.hg.1 NM_015057.4 Refseq
Affymetrix Human Clariom D Assay PSR1300162376.hg.1 ENST00000357337.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300162376.hg.1 ENST00000544440.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300162376.hg.1 NM_015057.4 Refseq
Affymetrix Human Clariom D Assay PSR1300162376.hg.1 TC1300009293.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0