This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01027910_m1
See other SLC30A9 GE Assays ›
Gene Symbol:
SLC30A9
Gene Name:
solute carrier family 30 member 9
Gene Aliases:
C4orf1, GAC63, HUEL, ZNT9
Chromosome Location:
Chr.4: 41990472 - 42087534 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01027910_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC30A9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006345.3 NP_006336.3 1 - 2 256 139
XM_017007654.1 XP_016863143.1 1 - 2 290 139
GenBank mRNA AF006621.2 - 1 - 2 230 139
AK225470.1 - 1 - 2 193 139
AK299917.1 - 1 - 2 232 139
AK312838.1 - 1 - 2 256 139
AY594282.1 - 1 - 2 110 139
BC000240.1 - 1 - 2 217 139
BC007732.2 - 1 - 2 217 4109702 139
BC016949.1 - 1 - 2 235 3850521 139
BG323207.1 - 256 139
KU178526.1 - 1 - 2 110 139
KU178527.1 - 1 - 2 110 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400007361.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000259.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK299917.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC30A9

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair
transcription, DNA-templated
zinc II ion transport
positive regulation of transcription from RNA polymerase II promoter
cation transmembrane transport
chromatin binding
transcription factor activity, sequence-specific DNA binding
cation transmembrane transporter activity
ligand-dependent nuclear receptor binding
ligand-dependent nuclear receptor transcription coactivator activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04003812.hg.1 ENST00000264451.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04003812.hg.1 ENST00000536881.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04003812.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04003812.hg.1 TC04000259.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04003813.hg.1 ENST00000264451.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04003813.hg.1 ENST00000536881.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04003813.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04003813.hg.1 TC04000259.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150355.hg.1 TC0400007361.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150354.hg.1 TC0400007361.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000505523.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000509683.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay TC0400007361.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 ENST00000264451.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 ENST00000510460.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 ENST00000513699.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 NM_006345.3 Refseq
Affymetrix Human Clariom D Assay PSR0400150356.hg.1 TC0400007361.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 123127 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0