Assay Details
Gene Symbol: ADCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174922.4 | NP_777582.4 | 8 - 9 | 1006 | 79 | |
XM_006716527.2 | XP_006716590.1 | 8 - 9 | 996 | 79 | ||
XM_011516907.2 | XP_011515209.1 | 8 - 9 | 1719 | 79 | ||
XM_011516909.1 | XP_011515211.1 | 8 - 9 | 1006 | 79 | ||
XM_011516910.1 | XP_011515212.1 | 9 - 10 | 1029 | 79 | ||
XM_017013174.1 | XP_016868663.1 | 8 - 9 | 1044 | 79 | ||
GenBank mRNA | AK092773.1 | - | 8 - 9 | 969 | 79 | |
BC085013.1 | - | 3 - 4 | 316 | 79 | ||
BC101659.1 | - | 8 - 9 | 946 | 8069165 | 79 | |
BC143326.1 | - | 7 - 8 | 773 | 79 | ||
BU845476.1 | - | 3 - 4 | 314 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000851.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08011360.hg.1 | ENST00000308860.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08011360.hg.1 | ENST00000526231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08011360.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08011360.hg.1 | TC08000851.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0800163414.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800163414.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800163414.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800163414.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800163414.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800163414.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800163414.hg.1 | TC0800009254.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
