This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01027002_m1
See other JAK1 GE Assays ›
Gene Symbol:
JAK1
Gene Name:
Janus kinase 1
Gene Aliases:
JAK1A, JAK1B, JTK3
Chromosome Location:
Chr.1: 64833223 - 65067746 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01027002_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: JAK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320923.1 NP_001307852.1 9 - 10 1399 81
NM_001321852.1 NP_001308781.1 8 - 9 1402 81
NM_001321853.1 NP_001308782.1 10 - 11 1661 81
NM_001321854.1 NP_001308783.1 9 - 10 1577 81
NM_001321855.1 NP_001308784.1 9 - 10 1580 81
NM_001321856.1 NP_001308785.1 8 - 9 1315 81
NM_001321857.1 NP_001308786.1 8 - 9 1496 81
NM_002227.3 NP_002218.2 8 - 9 1496 81
XM_017001212.1 XP_016856701.1 8 - 9 1343 81
GenBank mRNA AB209057.1 - 7 - 8 1252 81
AB219242.1 - 6 - 7 1146 81
AK314257.1 - 9 - 10 1401 81
BC132729.1 - 7 - 8 1214 40146751 81
HQ628631.1 - 6 - 7 1176 81
M64174.1 - 7 - 8 1221 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014457.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002741.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene JAK1

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
cell migration
cell differentiation
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
interleukin-2-mediated signaling pathway
regulation of cell proliferation
positive regulation of GTPase activity
innate immune response
response to antibiotic
interferon-gamma-mediated signaling pathway
regulation of interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
positive regulation of sprouting angiogenesis
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
growth hormone receptor binding
protein binding
ATP binding
protein phosphatase binding
ubiquitin protein ligase binding
CCR5 chemokine receptor binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000467764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000467764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000481702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000481702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000494904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000494904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000494904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0