Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
ENST00000395308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
ENST00000395309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
ENST00000395311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
ENST00000395314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
ENST00000413890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
ENST00000416613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
ENST00000438792.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
NM_001079521.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
NM_001079522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
NM_004986.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003513.hg.1 |
TC14000326.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395309.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395314.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000413890.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000438792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000459737.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_001079521.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_001079522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_001271014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_004986.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NR_073128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NR_073129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395309.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000395314.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000413890.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000438792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
ENST00000459737.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_001079521.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_001079522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_001271014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NM_004986.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NR_073128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
NR_073129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150211.hg.1 |
TC1400007244.hg.1 |
Clariom D TC |