Assay Details
Gene Symbol: ADGRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083909.2 | NP_001077378.1 | 2 - 3 | 340 | 102 | |
XM_017016779.1 | XP_016872268.1 | 1 - 2 | 200 | 102 | ||
GenBank mRNA | AK124728.1 | - | 2 - 3 | 340 | 102 | |
CB045391.1 | - | 2 - 3 | 334 | 102 | ||
DA208698.1 | - | 2 - 3 | 340 | 102 | ||
EF536972.1 | - | 1 - 2 | 129 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000948.hg.1
Additional Information:
This assay is in the 5'UTR region of the EF536972.1,NM_001083909.2,XM_017016779.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011705.hg.1 | ENST00000368577.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011705.hg.1 | ENST00000392607.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011705.hg.1 | ENST00000392609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011705.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011705.hg.1 | TC10000948.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10011701.hg.1 | ENST00000368577.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011701.hg.1 | ENST00000392607.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011701.hg.1 | ENST00000392609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011701.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011701.hg.1 | TC10000948.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10011700.hg.1 | ENST00000392607.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011700.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011700.hg.1 | TC10000948.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009444.hg.1 | NM_001291085.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164528.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164528.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164528.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164528.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164528.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164528.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164528.hg.1 | TC1000009444.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000164529.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164529.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164529.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164529.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164529.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164529.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164529.hg.1 | TC1000009444.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000164522.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164522.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164522.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164522.hg.1 | ENST00000392607.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164522.hg.1 | ENST00000607359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164522.hg.1 | NM_001083909.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164522.hg.1 | TC1000009444.hg.1 | Clariom D TC |
